missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant CMV CMV Pp52 Protein

A cDNA sequence encoding the CMV Pp52 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11464-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937348

  • £1100.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Pp52 Protein
Research Use Only
Recombinant Protein
E. coli
50mM Tris pH 7.2, 1mM EDTA and 50% glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
CMV
GST
CMV Pp52 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers