missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant CMV CMV Pp52 Protein

A cDNA sequence encoding the CMV Pp52 was constructed and used to recombinantly synthesize the protein.

£173.00 - £1100.00

Specifications

Name Pp52 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity CMV
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15927348
View Documents Promotion Details
enQuireBio™
QP11464-100UG
100 μg
£173.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15947348
View Documents Promotion Details
enQuireBio™
QP11464-500UG
500 μg
£660.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15937348
View Documents Promotion Details
enQuireBio™
QP11464-1MG
1 mg
£1100.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Pp52 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
CMV
GST
CMV Pp52 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
50mM Tris pH 7.2, 1mM EDTA and 50% glycerol.
Videos
SDS
Documents

Documents

Product Certifications