missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chlamydia W4 Protein

A cDNA sequence encoding the Chlamydia W4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11416-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927198

  • £173.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

W4 Protein
Research Use Only
Recombinant Protein
E. coli
(1 mg/1ml) 20mM Tris-HCl, pH 7.2, 50% glycerol and 1.5M urea.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Chlamydia
Untagged
Protein is >90% pure as determined by SDS PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers