missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia W4 Protein
A cDNA sequence encoding the Chlamydia W4 was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | W4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Chlamydia |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927198
|
enQuireBio™
QP11416-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947198
|
enQuireBio™
QP11416-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937198
|
enQuireBio™
QP11416-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
W4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
Untagged | |
Protein is >90% pure as determined by SDS PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/1ml) 20mM Tris-HCl, pH 7.2, 50% glycerol and 1.5M urea. |