missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Carp GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10631-10ug
Additional Details : Weight : 0.01000kg
Specifications
GH Protein | |
Research Use Only | |
Carp GH is biologically active in rat 3T3 F442A preadipocytes, though its activity is 15-fold lower compared to bovine GH, but it is equally potent in vivo in promoting carp growth (Fine et al. 1993). Furthermore, carp GH forms 1:2 complex with the extra cellular domain of ovine growth hormone receptor. | |
Carp | |
Untagged | |
The GH Carp was lyophilized from a concentrated (1 mg/ml) solution with 0.3% NaHCO3 adjusted to pH 8. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MARVLVLLSV VLVSLLVNQG RASDNQRLFN NAVIRVQHLH QLAAKMINDF EDSLLPEERR QLSKIFPLSF CNSDYIEAPA GKDETQKSSM LKLLRISFHL IESWEFPSQS LSGTVSNSLT VGNPNQLTEK LADLKMGISV LIQACLDGQP NMDDNDSLPL PFEDFYLTMG ENNLRESFRL LACFKKDMHK VETYLRVANC RRSLDSNCTL | |
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |