missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Carp GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
£99.15 - £2315.00
Specifications
Name | GH Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Carp GH is biologically active in rat 3T3 F442A preadipocytes, though its activity is 15-fold lower compared to bovine GH, but it is equally potent in vivo in promoting carp growth (Fine et al. 1993). Furthermore, carp GH forms 1:2 complex with the extra cellular domain of ovine growth hormone receptor. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944468
|
enQuireBio™
QP10631-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964468
|
enQuireBio™
QP10631-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954468
|
enQuireBio™
QP10631-1MG |
1 mg |
£2315.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GH Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MARVLVLLSV VLVSLLVNQG RASDNQRLFN NAVIRVQHLH QLAAKMINDF EDSLLPEERR QLSKIFPLSF CNSDYIEAPA GKDETQKSSM LKLLRISFHL IESWEFPSQS LSGTVSNSLT VGNPNQLTEK LADLKMGISV LIQACLDGQP NMDDNDSLPL PFEDFYLTMG ENNLRESFRL LACFKKDMHK VETYLRVANC RRSLDSNCTL | |
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Carp GH is biologically active in rat 3T3 F442A preadipocytes, though its activity is 15-fold lower compared to bovine GH, but it is equally potent in vivo in promoting carp growth (Fine et al. 1993). Furthermore, carp GH forms 1:2 complex with the extra cellular domain of ovine growth hormone receptor. | |
Carp | |
Untagged | |
The GH Carp was lyophilized from a concentrated (1 mg/ml) solution with 0.3% NaHCO3 adjusted to pH 8. |