missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine Leptin Protein
A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10754-100ug
Additional Details : Weight : 0.01000kg
Specifications
Leptin Protein | |
Research Use Only | |
Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. | |
Canine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02% NaHCO3. |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MRCGPLCRFL WLWPYLSCVE AVPIRKVQDD TKTLIKTIVA RINDISHTQS VSSKQRVAGL DFIPGLQPVL SLSRMDQTLA IYQQILNSLH SRNVVQISND LENLRDLLHL LASSKSCPLP RARGLETFES LGGVLEASLY STEVVALNRL QAALQDMLRR LDLSPGC | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |