missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine Leptin Protein
A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.
£99.15 - £1055.00
Specifications
Name | Leptin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965068
|
enQuireBio™
QP10754-20UG |
20 μg |
£99.15
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945068
|
enQuireBio™
QP10754-100UG |
100 μg |
£155.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955068
|
enQuireBio™
QP10754-1MG |
1 mg |
£1055.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Leptin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MRCGPLCRFL WLWPYLSCVE AVPIRKVQDD TKTLIKTIVA RINDISHTQS VSSKQRVAGL DFIPGLQPVL SLSRMDQTLA IYQQILNSLH SRNVVQISND LENLRDLLHL LASSKSCPLP RARGLETFES LGGVLEASLY STEVVALNRL QAALQDMLRR LDLSPGC | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. | |
Canine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02% NaHCO3. |