missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum CLOB-1 Protein
A DNA sequence encoding the Clostridium botulinum CLOB-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9572-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
CLOB-1 Protein | |
Research Use Only | |
HA-17 | |
Recombinant Protein | |
Yeast | |
SVERTFLPNG NYNIKSIFSG SLYLNPVSKS LTFSNESSAN NQKWNVEYMA ENRCFKISNV AEPNKYLSYD NFGFISLDSL SNRCYWFPIK IAVNTYIMLS LNKVNELDYA WDIYDTNENI LSQPLLLLPN FDIYNSNQMF KLEKI | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
C. botulinum | |
His | |
Tris-based buffer, 50% glycerol |