missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum CLOB-1 Protein

A DNA sequence encoding the Clostridium botulinum CLOB-1, was expressed in the hosts and tags indicated.

£321.05 - £2888.42

Specifications

Name CLOB-1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol HA-17
Protein Length Full Length
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16008491
View Documents Promotion Details
enQuireBio™
QP9572-YE-100UG
100 μg
£800.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16048491
View Documents Promotion Details
enQuireBio™
QP9572-YE-500UG
500 μg
£1930.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16018491
View Documents Promotion Details
enQuireBio™
QP9572-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16038491
View Documents Promotion Details
enQuireBio™
QP9572-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16058491
View Documents Promotion Details
enQuireBio™
QP9572-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16028491
View Documents Promotion Details
enQuireBio™
QP9572-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

CLOB-1 Protein
Not determined.
Full Length
C. botulinum
His
Tris-based buffer, 50% glycerol
Research Use Only
HA-17
Recombinant Protein
Yeast
SVERTFLPNG NYNIKSIFSG SLYLNPVSKS LTFSNESSAN NQKWNVEYMA ENRCFKISNV AEPNKYLSYD NFGFISLDSL SNRCYWFPIK IAVNTYIMLS LNKVNELDYA WDIYDTNENI LSQPLLLLPN FDIYNSNQMF KLEKI
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications