missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum CLOB-1 Protein

A DNA sequence encoding the Clostridium botulinum CLOB-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9572-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048491

  • £1930.00 / 500µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CLOB-1 Protein
Research Use Only
HA-17
Recombinant Protein
Yeast
SVERTFLPNG NYNIKSIFSG SLYLNPVSKS LTFSNESSAN NQKWNVEYMA ENRCFKISNV AEPNKYLSYD NFGFISLDSL SNRCYWFPIK IAVNTYIMLS LNKVNELDYA WDIYDTNENI LSQPLLLLPN FDIYNSNQMF KLEKI
Greater than 90% as determined by SDS-PAGE.
500 μg
Not determined.
Full Length
C. botulinum
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers