missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine MX1 Protein
A cDNA sequence encoding the MX1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | MX1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Bovine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961459
|
enQuireBio™
QP12761-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951459
|
enQuireBio™
QP12761-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941459
|
enQuireBio™
QP12761-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MX1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
His | |
The MX1 protein was lyophilized from a (1 mg/ml) 0.2microm filtered solution containing 20mM Tris-HCl, pH 7.9, 500mM NaCl, 0.5mM Imidazole, 0.1mM DTT and 6M urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHHSSGLVPRGSHMVHSDLGIEELDSPESSLNGSEDMESKSNLYSQYEEKVRPCIDLIDSLRSLGVEQDLALPAIAVIGDQSSGKSSVLEALSGVALPRGSGIVTRCPLVLRLKKLGNEDEWKGKVSFLDKEIEIPDASQVEKEISEAQIAIAGEGTGISHELISLEVSSPHVPDLTLIDLPGITRVAVGNQPPDIEYQIKSLIRKYILRQETINLVVVPANVDIATTEALRMAQEVDPQGDRTIGILTKPDLVDKGTEDKVVDVVRNLVFHLKKGYMIVKCRGQQDIKHRMSLDKALQRERIFFEDHAHFRDLLEEGKATIPCLAERLTSELIMHICKTLPLLENQIKETHQRITEELQKYGKDIPEEESEKMFCLIEKIDTFNKEIISTIEGEEFVEQYDSRLFTKVRAEFSKWSAVVEKNFEKGYEAIRKEIKQFENRYRGRELPGFVNYKTFETIIKKQVRVLEEPAVDMLHTVTDIIRNTFTDVSGKHFNEFFNLHRTAKSKIEDIRLEQENEAEKSIRLHFQMEQLVYCQDQVYRRALQQVREKEAEEEKNKKSNHYFQSQVSEPSTDEIFQHLTAYQQEVSTRISGHIPLIIQFFVLRTYGEQLKKSMLQLLQDKDQYDWLLKERTDTRDKRKFLKERLERLTRARQRLAKFPG | |
Greater than 90.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |