missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine MX1 Protein

A cDNA sequence encoding the MX1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12761-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951459

  • £155.00 / 20µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MX1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHHSSGLVPRGSHMVHSDLGIEELDSPESSLNGSEDMESKSNLYSQYEEKVRPCIDLIDSLRSLGVEQDLALPAIAVIGDQSSGKSSVLEALSGVALPRGSGIVTRCPLVLRLKKLGNEDEWKGKVSFLDKEIEIPDASQVEKEISEAQIAIAGEGTGISHELISLEVSSPHVPDLTLIDLPGITRVAVGNQPPDIEYQIKSLIRKYILRQETINLVVVPANVDIATTEALRMAQEVDPQGDRTIGILTKPDLVDKGTEDKVVDVVRNLVFHLKKGYMIVKCRGQQDIKHRMSLDKALQRERIFFEDHAHFRDLLEEGKATIPCLAERLTSELIMHICKTLPLLENQIKETHQRITEELQKYGKDIPEEESEKMFCLIEKIDTFNKEIISTIEGEEFVEQYDSRLFTKVRAEFSKWSAVVEKNFEKGYEAIRKEIKQFENRYRGRELPGFVNYKTFETIIKKQVRVLEEPAVDMLHTVTDIIRNTFTDVSGKHFNEFFNLHRTAKSKIEDIRLEQENEAEKSIRLHFQMEQLVYCQDQVYRRALQQVREKEAEEEKNKKSNHYFQSQVSEPSTDEIFQHLTAYQQEVSTRISGHIPLIIQFFVLRTYGEQLKKSMLQLLQDKDQYDWLLKERTDTRDKRKFLKERLERLTRARQRLAKFPG
Greater than 90.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Bovine
His
The MX1 protein was lyophilized from a (1 mg/ml) 0.2microm filtered solution containing 20mM Tris-HCl, pH 7.9, 500mM NaCl, 0.5mM Imidazole, 0.1mM DTT and 6M urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers