missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine HTF Protein

Recombinant Protein

Brand:  enQuireBio™ QP12351-1gram

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15980139

  • £770.00 / 1g
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

280705
1 g
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MRPAVRALLA CAVLGLCLAD PERTVRWCTI STHEANKCAS FRENVLRILE SGPFVSCVKK TSHMDCIKAI SNNEADAVTL DGGLVYEAGL KPNNLKPVVA EFHGTKDNPQ THYYAVAVVK KDTDFKLNEL RGKKSCHTGL GRSAGWNIPM AKLYKELPDP QESIQRAAAN FFSASCVPCA DQSSFPKLCQ LCAGKGTDKC ACSNHEPYFG YSGAFKCLME GAGDVAFVKH STVFDNLPNP EDRKNYELLC GDNTRKSVDD YQECYLAMVP SHAVVARTVG GKEDVIWELL NHAQEHFGKD KPDNFQLFQS PHGKDLLFKD SADGFLKIPS KMDFELYLGY EYVTALQNLR ESKPPDSSKD ECMVKWCAIG HQERTKCDRW SGFSGGAIEC ETAENTEECI AKIMKGEADA MSLDGGYLYI AGKCGLVPVL AENYKTEGES CKNTPEKGYL AVAVVKTSDA NINWNNLKDK KSCHTAVDRT AGWNIPMGLL YSKINNCKFD EFFSAGCAPG SPRNSSLCAL CIGSEKGTGK ECVPNSNERY YGYTGAFRCL VEKGDVAFVK DQTVIQNTDG NNNEAWAKNL KKENFEVLCK DGTRKPVTDA ENCHLARGPN HAVVSRKDKA TCVEKILNKQ QDDFGKSVTD CTSNFCLFQS NSKDLLFRDD TKCLASIAKK TYDSYLGDDY VRAMTNLRQC STSKLLEACT FHKP
Greater than 97.0% as determined by Cellulose Acetate.
HTF Protein
Research Use Only
TF
Bovine
Untagged
The protein (1 mg/ml) was lyophilized with no additives.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers