missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine HTF Protein
Recombinant Protein
£99.15 - £770.00
Specifications
Gene ID (Entrez) | 280705 |
---|---|
Name | HTF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | TF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990139
|
enQuireBio™
QP12351-50MG |
50 mg |
£99.15
50mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970139
|
enQuireBio™
QP12351-150MG |
150 mg |
£155.00
150mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980139
|
enQuireBio™
QP12351-1GRAM |
1 g |
£770.00
1g |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
280705 | |
Research Use Only | |
TF | |
Bovine | |
Untagged | |
The protein (1 mg/ml) was lyophilized with no additives. |
HTF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRPAVRALLA CAVLGLCLAD PERTVRWCTI STHEANKCAS FRENVLRILE SGPFVSCVKK TSHMDCIKAI SNNEADAVTL DGGLVYEAGL KPNNLKPVVA EFHGTKDNPQ THYYAVAVVK KDTDFKLNEL RGKKSCHTGL GRSAGWNIPM AKLYKELPDP QESIQRAAAN FFSASCVPCA DQSSFPKLCQ LCAGKGTDKC ACSNHEPYFG YSGAFKCLME GAGDVAFVKH STVFDNLPNP EDRKNYELLC GDNTRKSVDD YQECYLAMVP SHAVVARTVG GKEDVIWELL NHAQEHFGKD KPDNFQLFQS PHGKDLLFKD SADGFLKIPS KMDFELYLGY EYVTALQNLR ESKPPDSSKD ECMVKWCAIG HQERTKCDRW SGFSGGAIEC ETAENTEECI AKIMKGEADA MSLDGGYLYI AGKCGLVPVL AENYKTEGES CKNTPEKGYL AVAVVKTSDA NINWNNLKDK KSCHTAVDRT AGWNIPMGLL YSKINNCKFD EFFSAGCAPG SPRNSSLCAL CIGSEKGTGK ECVPNSNERY YGYTGAFRCL VEKGDVAFVK DQTVIQNTDG NNNEAWAKNL KKENFEVLCK DGTRKPVTDA ENCHLARGPN HAVVSRKDKA TCVEKILNKQ QDDFGKSVTD CTSNFCLFQS NSKDLLFRDD TKCLASIAKK TYDSYLGDDY VRAMTNLRQC STSKLLEACT FHKP | |
Greater than 97.0% as determined by Cellulose Acetate. |