missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Enterokinase Protein
A cDNA sequence encoding the Enterokinase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11793-EC-20IU
Additional Details : Weight : 0.00900kg
Specifications
Enterokinase Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSKRSVPSR HRSLTTYEVM FAVLFVILVA LCAGLIAVSW LSIQGSVKDA AFGKSHEARG TLKIISGATY NPHLQDKLSV DFKVLAFDIQ QMIDDIFQSS NLKNEYKNSR VLQFENGSII VIFDLLFDQW VSDKNVKEEL IQGIEANKSS QLVTFHIDLN SIDITASLEN FSTISPATTS EKLTTSIPLA TPGNVSIECP PDSRLCADAL KCIAIDLFCD GELNCPDGSD EDNKTCATAC DGRFLLTGSS GSFEALHYPK PSNNTSAVCR WIIRVNQGLS IQLNFDYFNT YYADVLNIYE GMGSSKILRA SLWSNNPGII RIFSNQVTAT FLIQSDESDY IGFKVTYTAF NSKELNNYEK INCNFEDGFC FWIQDLNDDN EWERTQGSTF PPSTGPTFDH TFGNESGFYI STPTGPGGRR ERVGLLTLPL DPTPEQACLS FWYYMYGENV YKLSINISSD QNMEKTIFQK EGNYGQNWNY GQVTLNETVE FKVSFYGFKN QILSDIALDD ISLTYGICNV SVYPEPTLVP TPPPELPTDC GGPHDLWEPN TTFTSINFPN SYPNQAFCIW NLNAQKGKNI QLHFQEFDLE NIADVVEIRD GEGDDSLFLA VYTGPGPVND VFSTTNRMTV LFITDNMLAK QGFKANFTTG YGLGIPEPCK EDNFQCKDGE CIPLVNLCDG FPHCKDGSDE AHCVRLFNGT TDSSGLVQFR IQSIWHVACA ENWTTQISDD VCQLLGLGTG NSSVPTFSTG GGPYVNLNTA PNGSLILTPS QQCLEDSLIL LQCNYKSCGK KLVTQEVSPK IVGGSDSREG AWPWVVALYF DDQQVCGASL VSRDWLVSAA HCVYGRNMEP SKWKAVLGLH MASNLTSPQI ETRLIDQIVI NPHYNKRRKN NDIAMMHLEM KVNYTDYIQP | |
Greater than 95.0% as determined by SDS-PAGE. |
20 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
Bovine EK in 50mM Tris-HCl, pH 8.0, 0.5M NaCl and 50% glycerol. |