missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Enterokinase Protein
A cDNA sequence encoding the Enterokinase was constructed and used to recombinantly synthesize the protein.
£109.00 - £1285.00
Specifications
Name | Enterokinase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Bovine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968388
|
enQuireBio™
QP11793-EC-20IU |
20 IU |
£109.00
20 units |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958388
|
enQuireBio™
QP11793-EC-100IU |
100 IU |
£393.00
100 units |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978388
|
enQuireBio™
QP11793-EC-500IU |
500 IU |
£1285.00
500 units |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Enterokinase Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
Bovine EK in 50mM Tris-HCl, pH 8.0, 0.5M NaCl and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSKRSVPSR HRSLTTYEVM FAVLFVILVA LCAGLIAVSW LSIQGSVKDA AFGKSHEARG TLKIISGATY NPHLQDKLSV DFKVLAFDIQ QMIDDIFQSS NLKNEYKNSR VLQFENGSII VIFDLLFDQW VSDKNVKEEL IQGIEANKSS QLVTFHIDLN SIDITASLEN FSTISPATTS EKLTTSIPLA TPGNVSIECP PDSRLCADAL KCIAIDLFCD GELNCPDGSD EDNKTCATAC DGRFLLTGSS GSFEALHYPK PSNNTSAVCR WIIRVNQGLS IQLNFDYFNT YYADVLNIYE GMGSSKILRA SLWSNNPGII RIFSNQVTAT FLIQSDESDY IGFKVTYTAF NSKELNNYEK INCNFEDGFC FWIQDLNDDN EWERTQGSTF PPSTGPTFDH TFGNESGFYI STPTGPGGRR ERVGLLTLPL DPTPEQACLS FWYYMYGENV YKLSINISSD QNMEKTIFQK EGNYGQNWNY GQVTLNETVE FKVSFYGFKN QILSDIALDD ISLTYGICNV SVYPEPTLVP TPPPELPTDC GGPHDLWEPN TTFTSINFPN SYPNQAFCIW NLNAQKGKNI QLHFQEFDLE NIADVVEIRD GEGDDSLFLA VYTGPGPVND VFSTTNRMTV LFITDNMLAK QGFKANFTTG YGLGIPEPCK EDNFQCKDGE CIPLVNLCDG FPHCKDGSDE AHCVRLFNGT TDSSGLVQFR IQSIWHVACA ENWTTQISDD VCQLLGLGTG NSSVPTFSTG GGPYVNLNTA PNGSLILTPS QQCLEDSLIL LQCNYKSCGK KLVTQEVSPK IVGGSDSREG AWPWVVALYF DDQQVCGASL VSRDWLVSAA HCVYGRNMEP SKWKAVLGLH MASNLTSPQI ETRLIDQIVI NPHYNKRRKN NDIAMMHLEM KVNYTDYIQP | |
Greater than 95.0% as determined by SDS-PAGE. |