missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacillus Levanase Protein
A DNA sequence encoding the Bacillus sp. (strain L7) Levanase, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Name | Levanase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Protein Length | Partial (see sequence information for more details). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048940
|
enQuireBio™
QP7478-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068940
|
enQuireBio™
QP7478-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058940
|
enQuireBio™
QP7478-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078940
|
enQuireBio™
QP7478-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088940
|
enQuireBio™
QP7478-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098940
|
enQuireBio™
QP7478-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Levanase Protein | |
Not determined. | |
Recombinant Protein | |
E. coli | |
LPWNDLGHVW SGSAVADTTN ASGLFGSSGG KGLIAYYTSY NPDRHNGNQK IGLAYSTDRG RTWKYSEEHP VVIENPGKTG EDPGGWDFRD PKVVRDEANN RWVMVVSGGD HIRLFTSTNL LNWTLTDQF | |
Greater than 90% as determined by SDS-PAGE. |
Research Use Only | |
Partial (see sequence information for more details). | |
Bacillus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |