missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant B. abortus Malate dehydrogenase Protein

A DNA sequence encoding the Brucella abortus (strain S19) Malate dehydrogenase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6351-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042670

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_002964995.1
500 μg
Not determined.
Full Length
B. abortus
His-SUMO
Tris-based buffer, 50% glycerol
Malate dehydrogenase Protein
Research Use Only
mdh
Recombinant Protein
E. coli
MARNKIALIG SGMIGGTLAH LAGLKELGDV VLFDIAEGTP QGKGLDIAES SPVDGFDAKF TGANDYAAIE GADVVIVTAG VPRKPGMSRD DLLGINLKVM EQVGAGIKKY APEAFVICIT NPLDAMVWAL QKFSGLPAHK VVGMAGVLDS ARFRYFLSEE FNVSVEDVTV FVLGGHGDSM VPLARYSTVA GIPLPDLVKM GWTSQDKLDK IIQRTRDGGA EIVGLLKTGS AFYAPAASAI QMAESYLKDK KRVLPVAAQL SGQYGVKDMY VGVPTVIGAN GVERIIEIDL DKDEKAQFDK SVASVAGLCE ACIGIAPSLK
Greater than 88.5% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers