missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant B. abortus Malate dehydrogenase Protein
A DNA sequence encoding the Brucella abortus (strain S19) Malate dehydrogenase, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | WP_002964995.1 |
---|---|
Name | Malate dehydrogenase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | mdh |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002670
|
enQuireBio™
QP6351-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022670
|
enQuireBio™
QP6351-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012670
|
enQuireBio™
QP6351-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032670
|
enQuireBio™
QP6351-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042670
|
enQuireBio™
QP6351-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052670
|
enQuireBio™
QP6351-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_002964995.1 | |
Research Use Only | |
mdh | |
Recombinant Protein | |
E. coli | |
MARNKIALIG SGMIGGTLAH LAGLKELGDV VLFDIAEGTP QGKGLDIAES SPVDGFDAKF TGANDYAAIE GADVVIVTAG VPRKPGMSRD DLLGINLKVM EQVGAGIKKY APEAFVICIT NPLDAMVWAL QKFSGLPAHK VVGMAGVLDS ARFRYFLSEE FNVSVEDVTV FVLGGHGDSM VPLARYSTVA GIPLPDLVKM GWTSQDKLDK IIQRTRDGGA EIVGLLKTGS AFYAPAASAI QMAESYLKDK KRVLPVAAQL SGQYGVKDMY VGVPTVIGAN GVERIIEIDL DKDEKAQFDK SVASVAGLCE ACIGIAPSLK | |
Greater than 88.5% as determined by SDS-PAGE. |
Malate dehydrogenase Protein | |
Not determined. | |
Full Length | |
B. abortus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |