missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ real-time PCR Tube Strips
GREENER_CHOICE

Product Code. 10046913 Shop All Eppendorf Products
Change view
Click to view available options
:
Quantity: 120/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10046913 Quantity: 120/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10046913 Supplier Eppendorf™ Supplier No. 0030132882
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Cap Style Without caps
Format 8 tubes in strip format for 0.1 mL
For Use With (Application) Real-Time PCR
No. per Pack 120/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.