missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Reagent Bottle, Wide Neck, Soda-Lime Glass, Amber, Standard Taper Ground Joint Neck
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Description
A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- UV protection due to external colouration
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- All glass components
- Tight closure due to standard ground joint
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents and chemicals; Storage of hygroscopic liquids, powders and granules |
| Includes | With standard ground glass flat-headed stopper |
| Material | Soda Lime Glass |
| Quantity | 10/Pk. |
| Diameter (Metric) | 86 mm |
| Autoclavable | Not Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction