missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Reagent Bottle, Wide Neck, Soda-Lime Glass, Amber, Standard Taper Ground Joint Neck

Product Code. 15211839 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15211839 100 mL Pack of 10
15221839 250 mL Pack of 10
15231839 500 mL Pack of 10
15241839 1000 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15211839 Supplier DWK Life Sciences Supplier No. 231882407
  • ONLINE EXCLUSIVE PRICE
    £182.70
    £178.47 / Pack of 10
    Save £4.23
    2% Off
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Classic wide-neck, glass-stoppered reagent bottles in amber soda lime glass.

A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!

  • UV protection due to external colouration
  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • All glass components
  • Tight closure due to standard ground joint

Specifications

Capacity (Metric) 100 mL
For Use With (Application) Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents and chemicals; Storage of hygroscopic liquids, powders and granules
Includes With standard ground glass flat-headed stopper
Material Soda Lime Glass
Quantity 10/Pk.
Diameter (Metric) 52 mm
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.