missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Reagent Bottle, Narrow Neck, Soda-Lime Glass, Amber, Standard Taper Ground Joint Neck
Classic narrow-neck, glass-stoppered reagent bottles in amber soda lime glass.
£138.00 - £213.00
Specifications
For Use With (Application) | Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents and chemicals; Storage of hygroscopic liquids, powders and granules |
---|---|
Includes | With standard ground glass flat-headed stopper |
Material | Soda Lime Glass |
Quantity | 10/Pk. |
Autoclavable | Not Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15221819
|
DWK Life Sciences
231681706 |
50 mL |
£138.00
Pack of 10 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
15231819
|
DWK Life Sciences
231682402 |
100 mL |
£141.00
Pack of 10 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
15251819
|
DWK Life Sciences
231684403 |
500 mL |
£187.00
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
15241819
|
DWK Life Sciences
231683604 |
250 mL |
£146.00
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
15261819
|
DWK Life Sciences
231685408 |
1000 mL |
£213.00
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
Description
A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- UV protection due to external colouration
- All glass components
- Tight closure due to standard ground joint
Specifications
Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents and chemicals; Storage of hygroscopic liquids, powders and granules | |
Soda Lime Glass | |
Not Autoclavable |
With standard ground glass flat-headed stopper | |
10/Pk. |