missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Reagent Borosilicate Glass Bottle with Cap

Product Code. 12997640 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12997640 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12997640
Discount Lab Supplies
Use promo code 28170
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Measure laboratory media using the Fisherbrand™ Reagent Borosilicate Glass Bottle with Cap. It conforms to ISO R1115 and DIN 168, is chemically resistant and can be repeatedly sterilized to 140°C.

Use these glass media bottles for a variety of general laboratory applications. Fisherbrand™ Reusable Glass Media Bottles with Cap feature a convenient wide opening, enhanced graduations and a polypropylene pour ring. Feature clear (natural), drip-f

  • 20,000mL
  • Graduated Blue polypropylene cap
  • Pouring ring
  • 45 screw thread standard
  • Conforms to ISO R1115 and DIN 168
  • Not autoclavable with tightly screwed cap

Specifications

Capacity 676.3 oz., 20000 mL
Capacity (Metric) 20,000 mL
Color Clear
Diameter (English) Inner 1.18 in.
Diameter (English) 11.3 in.
Diameter (Metric) Inner Neck 30 mm
For Use With (Application) Media and Reagent Storage, Holding and Dispensing Fluids, Chemical Storage
Includes Screw Cap, Pouring Ring
Material Borosilicate Glass (3.3)
No. per Pack 10 Pack
Quantity 1 Ea.
Sterility Sterile
Certified Yes
Closure Color Blue
Graduations Yes
Neck Finish Narrow
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 505 mm
Height (English) 19.88 in.
Chemical Label Unlabeled
Closure Material Polypropylene
Closure Size GL 45
Diameter (Metric) 288 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 676.3 oz.
Shape Cylindrical
Certifications/Compliance ISO 4796
Disposable Reusable
Product Type McCartney Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.