missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyLabel™ Antibody Labeling Kits

Product Code. 18785455
Change view
Click to view available options
Quantity:
5 x 100 μg of IgG
5 x 20 μg
5 x 20 μg of IgG
Label or Dye:
Alexa Fluor 488
Alexa Fluor 514
Alexa Fluor 594
Alexa Fluor 647
Alexa Fluor 700
Alexa Fluor Plus 555
Alexa Fluor Plus 750
Biotin
None
Unit Size:
5 reactions
Pack of 5
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Label or Dye unitSize
18785455 5 x 20 μg of IgG Alexa Fluor 488 5 reactions
18725475 5 x 100 μg of IgG Alexa Fluor 488 5 reactions
17903101 5 x 20 μg of IgG Alexa Fluor 647 Pack of 5
18755435 5 x 100 μg of IgG Alexa Fluor 647 5 reactions
18745435 5 x 20 μg of IgG Biotin 5 reactions
18785385 5 x 100 μg of IgG Biotin 5 reactions
18041683 5 x 20 μg of IgG None 5 reactions
18041663 5 x 100 μg of IgG None 5 reactions
30245080 5 x 20 μg Alexa Fluor 514 Pack of 5
30245274 5 x 20 μg Alexa Fluor Plus 555 Pack of 5
30245024 5 x 20 μg Alexa Fluor 594 Pack of 5
30245137 5 x 20 μg Alexa Fluor 700 Pack of 5
30245465 5 x 20 μg Alexa Fluor Plus 750 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 18785455 Supplier Invitrogen™ Supplier No. R10709
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ReadyLabel Antibody Labeling Kits enable fast, convenient labeling of off-the-shelf antibodies and cell culture supernatants without prior purification of the antibody solution.

ReadyLabel Antibody Labeling Kits enable fast, convenient labeling of off-the-shelf antibodies and cell culture supernatants without prior purification of the antibody solution. Each kit contains everything needed to perform 5 labeling reactions and will produce purified antibody conjugates, without the purification step.

Each ReadyLabel Antibody Labeling Kit, 100 μg contains everything needed to perform 5 labeling reactions and will produce purified antibody conjugates, without the purification step. The kit can be used to label antibodies in solutions containing BSA or other stabilizing proteins and antibodies in a primary amine-containing buffer such as Tris or glycine, or even cell culture supernatants, without prior purification of the antibody-containing solution. A reactive dye moiety efficiently binds to primary amines of antibodies to form stable, covalently bound antibody-dye conjugates.

Alexa Fluor dyes produce antibody conjugates that are typically brighter with better photostability and lower pH sensitivity relative to traditional dyes (e.g., FITC). Unlike other dyes, the fluorescence of Alexa Fluor conjugates are insensitive to pH between 4 and 10. Alexa Fluor-labeled antibodies can be used for applications including microscopy, flow cytometry, western blotting, ELISA, and indirect FISH.

ReadyLabel Antibody Labeling Kits offer the following benefits:

  • Convenience—Labeling takes one hour, with less than 20 minutes of hands-on time
  • Versatility—Labelling can be performed on IgG antibodies, regardless of species, isotype, or storage buffer
  • Customization—ReadyLabel Flex Antibody Labeling Kits are designed for use with a wide range of amine-reactive labels including Alexa Fluor dyes
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Production and Labeling
Product Type Antibody Labeling Kit
Quantity 5 x 20 μg of IgG
For Use With (Equipment) Flow Cytometer, Fluorescence Microscope
Product Line ReadyLabel
Chemical Reactivity Amine
Solubility DMSO (Dimethylsulfoxide)
Label or Dye Alexa Fluor 488
Color Green
Do rat or bovine antibodies bind to the ReadyLabel columns?

ReadyLabel Columns bind poorly to rat and bovine antibodies, and are therefore, not recommended to use with the ReadyLabel Antibody Labeling Kits (Cat. Nos. R10701, R10702, R10709, R10710, R10711, R10712, R10713, R10714).

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.