missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | RDH5 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RDH5 Polyclonal antibody specifically detects RDH5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| RDH5 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 5959 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 11-cis RDH, 11-cis retinol dehydrogenase, 9,9-cis-retinol specific dehydrogenase, EC 1.1.1, EC 1.1.1.105, FLJ39337, FLJ97089, HSD17B, RDH1, retinol dehydrogenase 1, retinol dehydrogenase 5 (11-cis and 9-cis), retinol dehydrogenase 5 (11-cis/9-cis), SDR9C5, short chain dehydrogenase/reductase family 9C, member 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-318 of human RDH5 (NP_002896.2). LPASNAFVFITGCDSGFGRLLALQLDQRGFRVLASCLTPSGAEDLQRVASSRLHTTLLDITDPQSVQQAAKWVEMHVKEAGLFGLVNNAGVAGIIGPTPWLTRDDFQRVLNVNTMGPIGVTLALLPLLQQARGRVINITSVLGRLAANGGGYCVSKFGLEAFSDSLRRDVAHFGIRVSIVEPGFFRTPVTNLESLEKTLQACWARLPPATQAHYGGAFLTKYLKMQQRIMNLICDPDLTKVSRCLEHALTARHPRTRYSPGWDAKLLWLPASYLPASLVDAVLTWVLPKPAQAVY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title