missing translation for 'onlineSavingsMsg'
Learn More

RDH5 Antibody - BSA Free, Novus Biologicals™

Product Code. 18649220 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18649220 0.1 mL 0.01mL
18691641 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18649220 Supplier Novus Biologicals Supplier No. NBP2937540.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RDH5 Polyclonal antibody specifically detects RDH5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH5
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias 11-cis RDH, 11-cis retinol dehydrogenase, 9,9-cis-retinol specific dehydrogenase, EC 1.1.1, EC 1.1.1.105, FLJ39337, FLJ97089, HSD17B, RDH1, retinol dehydrogenase 1, retinol dehydrogenase 5 (11-cis and 9-cis), retinol dehydrogenase 5 (11-cis/9-cis), SDR9C5, short chain dehydrogenase/reductase family 9C, member 5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-318 of human RDH5 (NP_002896.2). LPASNAFVFITGCDSGFGRLLALQLDQRGFRVLASCLTPSGAEDLQRVASSRLHTTLLDITDPQSVQQAAKWVEMHVKEAGLFGLVNNAGVAGIIGPTPWLTRDDFQRVLNVNTMGPIGVTLALLPLLQQARGRVINITSVLGRLAANGGGYCVSKFGLEAFSDSLRRDVAHFGIRVSIVEPGFFRTPVTNLESLEKTLQACWARLPPATQAHYGGAFLTKYLKMQQRIMNLICDPDLTKVSRCLEHALTARHPRTRYSPGWDAKLLWLPASYLPASLVDAVLTWVLPKPAQAVY
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5959
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.