missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH11 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£166.00 - £305.00
Specifications
| Antigen | RDH11 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:10 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:2000, Immunohistochemistry-Frozen 1:10 - 1:2000 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18139873
|
Novus Biologicals
NBP2-27584SS |
0.025 mg |
£166.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18119053
|
Novus Biologicals
NBP2-27584 |
0.1 mg |
£305.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RDH11 Polyclonal specifically detects RDH11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| RDH11 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:10 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:2000, Immunohistochemistry-Frozen 1:10 - 1:2000 | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| PBS with 0.02% Sodium Azide | |
| Androgen-regulated short-chain dehydrogenase/reductase 1, ARSDR1CGI82, EC 1.1.1.300, HCBP12, HCV core-binding protein HCBP12, MDT1, prostate short-chain dehydrogenase reductase 1, Prostate short-chain dehydrogenase/reductase 1, PSDR1FLJ32633, RALR1, Retinal reductase 1, retinol dehydrogenase 11, retinol dehydrogenase 11 (all-trans and 9-cis), retinol dehydrogenase 11 (all-trans/9-cis/11-cis), SCALD, SDR7C1, short chain dehydrogenase/reductase family 7C, member 1 | |
| RDH11 | |
| IgG | |
| Affinity Purified | |
| Reacts with Human. RDH11 protein of Human is 86% identical with Mouse protein. |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8TC12 | |
| 51109 | |
| Synthetic peptides corresponding to the N terminal sequence MVELMFP and the C terminal sequence LLGLPID of the human RDH11 PSDR1 protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 35.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title