missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH11 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27584
This item is not returnable.
View return policy
Description
RDH11 Polyclonal specifically detects RDH11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| RDH11 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Androgen-regulated short-chain dehydrogenase/reductase 1, ARSDR1CGI82, EC 1.1.1.300, HCBP12, HCV core-binding protein HCBP12, MDT1, prostate short-chain dehydrogenase reductase 1, Prostate short-chain dehydrogenase/reductase 1, PSDR1FLJ32633, RALR1, Retinal reductase 1, retinol dehydrogenase 11, retinol dehydrogenase 11 (all-trans and 9-cis), retinol dehydrogenase 11 (all-trans/9-cis/11-cis), SCALD, SDR7C1, short chain dehydrogenase/reductase family 7C, member 1 | |
| Rabbit | |
| 35.4 kDa | |
| 0.1 mg | |
| Vision | |
| 51109 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:10 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:2000, Immunohistochemistry-Frozen 1:10 - 1:2000 | |
| Q8TC12 | |
| RDH11 | |
| Synthetic peptides corresponding to the N terminal sequence MVELMFP and the C terminal sequence LLGLPID of the human RDH11 PSDR1 protein | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with Human. RDH11 protein of Human is 86% identical with Mouse protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction