missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ VACUUBRAND™ RC6 Chemistry-HYBRID Vacuum Pump

Product Code. 10693562 Shop All Vacuubrand Products
Click to view available options
Plug Type:
CEE
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10693562

Brand: BrandTech™ VACUUBRAND™ 20698560

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For fine-vacuum applications

Combines rotary vane and diaphragm vacuum pump technology to support freeze drying and molecular distillations, with substantially reduced maintenance and less hazardous oil waste. Chemistry-design diaphragm pump is used to back the rotary vane pump. This configuration keeps pump oil under vacuum, reducing condensation of solvent vapors in pump oil and oil changes by as much as 90% over conventional rotary vane designs.

  • Free air capacity: 4.1cfm
  • Ultimate vacuum: 4 x 10-4mbar
  • Restriction-free, high-flow inlet ensures high flowrates under vacuum
  • Includes integrated oil mist filter

Order Info

Dimensions: 20L x 12W x9 in. H (51 x 30.5 x 23cm). Net weight: 53 lb. (24.2kg).

TRUSTED_SUSTAINABILITY

Specifications

Horsepower 0.5 hp
Amperage 2.6/2.7 A
Includes GKF 1000i cold trap, Oil mist filter, valve, T-connection, gas ballast
Motor Speed 1500 to 1800
Certifications/Compliance UL
Electrical Requirements 230 V 50/60 Hz
Model RC6
Noise Level 50 dbA
Plug Type CEE
Temperature (Metric) Storage -10°C to +60°C
Weight (English) 53.3 lb.
Weight (Metric) 24.2 kg
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 12°C
Flow Rate 3.5/4.1 CFM
For Use With Freeze dryers, benchtop centrifugal concentrators or other low-temperature evaporations of high boiling point solvents, molecular distillations
Frequency 50/60 Hz
Overall Height (English) 9 in.
Overall Height (Metric) 23 cm
Overall Length (English) 20 in.
Overall Length (Metric) 51 cm
Overall Width (English) 12 in.
Enclosure Rating IP40
Maximum Pressure 1.1 bar
Voltage 230 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.