missing translation for 'onlineSavingsMsg'
Learn More

RBM46 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-80475

 View more versions of this product

Product Code. 18258565

  • £381.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RBM46 Polyclonal specifically detects RBM46 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RBM46
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
Cancer/testis antigen 68, CT68MGC27016, probable RNA-binding protein 46, RNA binding motif protein 46, RNA-binding motif protein 46
Rabbit
Affinity purified
RUO
166863
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.5 mg/ml
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
NP_659416
RBM46
Synthetic peptide directed towards the N terminal of human MGC27016. Peptide sequence LNNYEIRPGKFIGVCVSLDNCRLFIGAIPKEKKKEEILDEMKKVTEGVVD.
100 μL
Primary
Expected identity based on immunogen sequence: Rat: 100%; Bovine: 100%; Canine: 100%; Mouse: 100%; Crab-eating macaque: 100%; Chicken: 85%; African malaria mosquito: 84%; Yellowfever mosquito: 84%; Harpegnathos saltator: 84%; Zebrafish: 76%; Western clawed frog: 76%; Florida lancelet: 76%; Green puffer: 76%; Slime mold: 76%; Sumatran orangutan: 76%; Human: 100%.
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.