missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM15 Mouse anti-Human, CoraLite 488, Clone: 4A1A4, Proteintech
Mouse Monoclonal Antibody
£128.70 - £319.04
Specifications
| Antigen | RBM15 |
|---|---|
| Clone | 4A1A4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | CoraLite 488 |
Description
RBM15 Monoclonal antibody apecifically detetects RBM15 antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| RBM15 | |
| Flow Cytometry | |
| CoraLite 488 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 64783 | |
| RBM15 fusion protein Ag17970 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 4A1A4 | |
| Monoclonal | |
| Liquid | |
| Human | |
| BC006397 | |
| RBM15 | |
| IgG2a | |
| Protein A purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title