missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM15 Mouse anti-Human, CoraLite 488, Clone: 4A1A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66059-100UL
This item is not returnable.
View return policy
Description
RBM15 Monoclonal antibody apecifically detetects RBM15 antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| RBM15 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| RBM15 | |
| RBM15 fusion protein Ag17970 | |
| 100 μL | |
| 64783 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG2a |
| Flow Cytometry | |
| 4A1A4 | |
| CoraLite 488 | |
| BC006397 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction