missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP9 Mouse anti-Human, Clone: 3D5E11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RBBP9 |
|---|---|
| Clone | 3D5E11 |
| Concentration | 0.81 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16837515
|
Proteintech
66015-2-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16827515
|
Proteintech
66015-2-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.Specifications
| RBBP9 | |
| 0.81 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BOG, RBBP9 | |
| RBBP9 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 3D5E11 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75884 | |
| 10741 | |
| RBBP9 Fusion Protein Ag17864 | |
| Primary | |
| -20°C | |
| RBBP9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title