missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP9 Mouse anti-Human, Clone: 3D5E11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66015-2-IG-150UL
This item is not returnable.
View return policy
Description
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.Specifications
| RBBP9 | |
| Monoclonal | |
| 0.81 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75884 | |
| RBBP9 | |
| RBBP9 Fusion Protein Ag17864 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3D5E11 | |
| Unconjugated | |
| RBBP9 | |
| BOG, RBBP9 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10741 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction