missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAVER2 Recombinant Rabbit Monoclonal Antibody (24GB810)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557678
This item is not returnable.
View return policy
Description
RAVER2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
RAVER2 may bind single-stranded nucleic acids.
Specifications
| RAVER2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RAVER2 | |
| A430091O22Rik; KIAA1579; Protein raver-2; RAVER2; Ribonucleoprotein PTB-binding 2; ribonucleoprotein, PTB binding 2; ribonucleoprotein, PTB-binding 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55225 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB810 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HCJ3 | |
| RAVER2 | |
| A synthesized peptide derived from human RAVER2 (aa 481-691). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction