missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAVER2 Recombinant Rabbit Monoclonal Antibody (24GB810)
Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | RAVER2 |
|---|---|
| Clone | 24GB810 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30276493
|
Invitrogen™
MA557678 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275555
|
Invitrogen™
MA557679 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAVER2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
RAVER2 may bind single-stranded nucleic acids.Specifications
| RAVER2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| RAVER2 | |
| A430091O22Rik; KIAA1579; Protein raver-2; RAVER2; Ribonucleoprotein PTB-binding 2; ribonucleoprotein, PTB binding 2; ribonucleoprotein, PTB-binding 2 | |
| RAVER2 | |
| Primary | |
| Affinity Chromatography |
| 24GB810 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HCJ3 | |
| 55225 | |
| A synthesized peptide derived from human RAVER2 (aa 481-691). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title