missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Rat Nogo-A Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £351.00
Specifikationer
| Antigen | Nogo-A |
|---|---|
| Clone | 391401 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
18763778
![]() |
R&D Systems
MAB30981-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18168840
![]() |
R&D Systems
MAB30981 |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
Nogo-A Monoclonal specifically detects Nogo-A in Rat samples. It is validated for Western Blot.Specifikationer
| Nogo-A | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat | |
| Q9JK11 | |
| Rtn4 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 391401 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 57142 | |
| E. coli-derived recombinant rat Nogo-A Glu2-Val172 Accession # Q9JK11 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat Nogo-A in direct ELISAs and Western blots. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel