missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Nogo-A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB30981
This item is not returnable.
View return policy
Description
Nogo-A Monoclonal specifically detects Nogo-A in Rat samples. It is validated for Western Blot.
Specifications
| Nogo-A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9JK11 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57142 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 391401 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Rtn4 | |
| E. coli-derived recombinant rat Nogo-A Glu2-Val172 Accession # Q9JK11 | |
| 100 μg | |
| Primary | |
| Detects rat Nogo-A in direct ELISAs and Western blots. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction