Learn More
Rat IgG2a kappa Isotype Control (eBR2a), Functional Grade, eBioscience™, Invitrogen™
Rat Isotype Control
£48.09 - £151.00
Specifications
Antigen | Rat IgG2a kappa |
---|---|
Clone | eBR2a |
Concentration | 1 mg/mL |
Applications | Control, Flow Cytometry, Functional Assay |
Conjugate | Functional Grade |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15248617
|
Affymetrix eBioscience
16-4321-81 |
50 μg |
£48.09
50µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15258617
|
Affymetrix eBioscience
16-4321-82 |
100 μg |
£83.00
100µg |
In Stock
Log in to see stock. |
|||||
15268617
|
Affymetrix eBioscience
16-4321-85 |
500 μg |
£151.00
500µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2a isotype control has been reported for use in flow cytometric analysis and ELISA. Applications Tested: This rat IgG2a isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and can be used at the same concentration as the experimental antibody. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.Specifications
Rat IgG2a kappa | |
1 mg/mL | |
Functional Grade | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C |
eBR2a | |
Control, Flow Cytometry, Functional Assay | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.