missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2a kappa Isotype Control (eBR2a), Functional Grade, eBioscience™, Invitrogen™

Rat Isotype Control

Brand:  Affymetrix eBioscience 16-4321-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15248617

  • £48.09 / 50µg
Estimated Shipment: 03-05-2024
to see stock.
Add to basket

Description

Description

Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2a isotype control has been reported for use in flow cytometric analysis and ELISA. Applications Tested: This rat IgG2a isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and can be used at the same concentration as the experimental antibody. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.
Specifications

Specifications

Rat IgG2a kappa
eBR2a
Functional Grade
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry, Functional Assay
1 mg/mL
PBS with no preservative; pH 7.2
Rat
50 μg
Isotype Control (Primary)
4° C
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.