missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CCR2 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
£223.00 - £403.00
Specifications
| Antigen | CCR2 |
|---|---|
| Clone | 890231 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18777098
|
R&D Systems
FAB8368V-100UG |
100 μg |
£403.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18735628
|
R&D Systems
FAB8368V-025UG |
25 μg |
£223.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR2 Monoclonal specifically detects CCR2 in Rat samples. It is validated for Flow Cytometry.Specifications
| CCR2 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Mouse | |
| Rat | |
| O55193 | |
| 729230 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant |
| 890231 | |
| Flow Cytometry | |
| Alexa Fluor 405 | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| CCR2 | |
| Primary | |
| Stains rat CCR2 transfectants but not irrelevant transfectants in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title