missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CCR2 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB8368V-100UG
This item is not returnable.
View return policy
Description
CCR2 Monoclonal specifically detects CCR2 in Rat samples. It is validated for Flow Cytometry.
Specifications
| CCR2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1.0 μg/106 cells | |
| O55193 | |
| CCR2 | |
| NS0 mouse myeloma cell line transfected with rat CCR2 Accession Number- O55193 | |
| 100 μg | |
| Primary | |
| Stains rat CCR2 transfectants but not irrelevant transfectants in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. |
| Flow Cytometry | |
| 890231 | |
| Alexa Fluor 405 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 729230 | |
| Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction