missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgM Heavy Chain Secondary Antibody, FITC, Novus Biologicals™
Rat Monoclonal Secondary Antibody
£228.00 - £317.00
Specifications
| Antigen | IgM |
|---|---|
| Clone | eB121-15F9 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274562
|
Novus Biologicals
NBP1-43944-0.05MG |
0.05 mg |
£228.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18288483
|
Novus Biologicals
NBP1-43944-0.1MG |
0.1 mg |
£270.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18234122
|
Novus Biologicals
NBP1-43944-0.5MG |
0.5 mg |
£317.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgM Heavy Chain Monoclonal specifically detects IgM Heavy Chain in Mouse samples. It is validated for Flow Cytometry.Specifications
| IgM | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Mouse | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| IGHM | |
| IgG2a κ | |
| Affinity Purified |
| eB121-15F9 | |
| Flow Cytometry | |
| Monoclonal | |
| Rat | |
| Immunology | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
| 3507 | |
| The immunogen for this antibody was IgM. | |
| Secondary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title