missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgM Heavy Chain Secondary Antibody, FITC, Novus Biologicals™

Rat Monoclonal Secondary Antibody

Brand:  Novus Biologicals NBP1-43944-0.1MG

214.20 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 18288483

  • £270.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgM Heavy Chain Monoclonal specifically detects IgM Heavy Chain in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgM
Monoclonal
0.5 mg/mL
Flow Cytometry
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Rat
Affinity Purified
RUO
Secondary
Mouse
IgG2a κ
Flow Cytometry
eB121-15F9
FITC
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
IGHM
The immunogen for this antibody was IgM.
0.1 mg
Immunology
3507
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only