missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRP3 Mouse anti-Human, Mouse, Clone: 1A10G4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RASGRP3 |
|---|---|
| Clone | 1A10G4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868725
|
Proteintech
66650-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858725
|
Proteintech
66650-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Members of the RAS.Specifications
| RASGRP3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| GRP3, KIAA0846, Ras guanyl releasing protein 3, RASGRP3 | |
| Rasgrp3 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1A10G4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| Q8IV61 | |
| 240168, 25780 | |
| RASGRP3 Fusion Protein Ag3810 | |
| Primary | |
| -20°C | |
| RASGRP3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title