missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRP3 Mouse anti-Human, Mouse, Clone: 1A10G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66650-1-IG-20UL
This item is not returnable.
View return policy
Description
Members of the RAS.Specifications
| RASGRP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IV61 | |
| Rasgrp3 | |
| RASGRP3 Fusion Protein Ag3810 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1A10G4 | |
| Unconjugated | |
| RASGRP3 | |
| GRP3, KIAA0846, Ras guanyl releasing protein 3, RASGRP3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 240168, 25780 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction