missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASEF Recombinant Rabbit Monoclonal Antibody (23GB2745), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | RASEF |
|---|---|
| Clone | 23GB2745 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088527
![]() |
Invitrogen
MA552520 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089717
|
Invitrogen
MA552521 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RASEF Recombinant Monoclonal antibody specifically detects RASEF in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| RASEF | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q5RI75, Q8IZ41 | |
| 158158, 242505, 298138 | |
| IgG | |
| Affinity Chromatography | |
| Antibody |
| 23GB2745 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BC030882; RAB45; RAS and EF hand domain containing; RAS and EF-hand domain containing; ras and EF-hand domain-containing protein; Ras and EF-hand domain-containing protein homolog; RASEF; ras-related protein Rab-45 | |
| A synthesized peptide derived from human RAB45 (82-120AA). | |
| Primary | |
| -20°C | |
| RASEF |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title