missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASEF Recombinant Rabbit Monoclonal Antibody (23GB2745), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA552520
This item is not returnable.
View return policy
Description
RASEF Recombinant Monoclonal antibody specifically detects RASEF in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| RASEF | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RASEF | |
| BC030882; RAB45; RAS and EF hand domain containing; RAS and EF-hand domain containing; ras and EF-hand domain-containing protein; Ras and EF-hand domain-containing protein homolog; RASEF; ras-related protein Rab-45 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 158158, 242505, 298138 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2745 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5RI75, Q8IZ41 | |
| RASEF | |
| A synthesized peptide derived from human RAB45 (82-120AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction